Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-6-chloropyridazine-3-carboxylate 98%

Supplier: AMBEED, INC
Warning

A163369-1G A163369-5G A163369-10G A163369-1G A163369-25G A163369-5G A163369-100G
103805-720EA 15 USD InStock
103805-720 103843-980 76720-218 77320-070 77320-072 77320-074 77879-104
Methyl-6-chloropyridazine-3-carboxylate 98%
Methyl-6-chloropyridazine-3-carboxylate
Precaución: for Research Use Only


Formula: C₆H₅ClN₂O₂
Boiling Pt: 310.9 °C (760 mm Hg)
Flash Pt: 141.851 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08694876
CAS Number: 65202-50-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty