Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-6-formylnicotinate 97%

Supplier: AMBEED, INC
Warning

A157975-1G A157975-5G A157975-100MG A157975-250MG A157975-25G A157975-5G
103805-074EA 61 USD InStock
103805-074 103843-280 76720-002 76720-004 77319-122 77319-124
Methyl-6-formylnicotinate 97%
Methyl-6-formylnicotinate
Precaución: for Research Use Only


Formula: C₈H₇NO₃
Boiling Pt: 272.9 °C (760 mm Hg)
Melting Pt: 118…120 °C
Flash Pt: 119 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08275010
CAS Number: 10165-86-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty