Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 8-(tert-butyl)-6-chloroimidazo[1,2-b]pyridazine-2-carboxylate 97%

Supplier: AMBEED, INC
Warning

A261994-1G A261994-5G A261994-100MG A261994-250MG A261994-1G A261994-5G
103812-552EA 393 USD InStock
103812-552 103852-444 77238-526 77238-528 77304-840 77304-842
Methyl 8-(tert-butyl)-6-chloroimidazo[1,2-b]pyridazine-2-carboxylate 97%
Methyl 8-(tert-butyl)-6-chloroimidazo[1,2-b]pyridazine-2-carboxylate
Precaución: for Research Use Only


Formula: C₁₂H₁₄ClN₃O₂
MW: 267.71 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD28144723
CAS Number: 1690176-75-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty