Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-9-hydroxynonanoate ≥95%

Supplier: AMBEED, INC
Warning

A128201-1G A128201-100MG A128201-250MG
103799-328EA 10 USD InStock
103799-328 103801-088 77624-086 77624-088
Methyl-9-hydroxynonanoate ≥95%
Methyl-9-hydroxynonanoate
Precaución: Sealed in dry, at 2…8°C


Formula: C₁₀H₂₀O₃
Storage Temperature: Refrigerator
MDL Number: MFCD00191518
CAS Number: 34957-73-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty