Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl imidazo[1,5-a]pyridine-8-carboxylate 98%

Supplier: AMBEED, INC
Warning

A128218-1G A128218-100MG A128218-250MG A128218-1G
103801-090EA 391 USD InStock
103801-090 76822-822 76822-824 77298-274
Methyl imidazo[1,5-a]pyridine-8-carboxylate 98%
Methyl imidazo[1,5-a]pyridine-8-carboxylate
Precaución: for Research Use Only


Formula: C₉H₈N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD21336569
CAS Number: 151509-02-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty