Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ac-Phe-OMe 97%

Supplier: AMBEED, INC
Warning

A148166-1G A148166-5G A148166-100G A148166-10G A148166-25G A148166-1G A148166-5G
103803-884EA 6 USD InStock
103803-884 103841-998 76689-250 76689-252 76689-254 77299-644 77317-432
Ac-Phe-OMe 97%
Ac-Phe-OMe
Precaución: for Research Use Only


Formula: C₁₂H₁₅NO₃
Boiling Pt: 391 °C (760 mm Hg)
Flash Pt: 190.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00066060
CAS Number: 3618-96-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty