Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl oxazole-2-carboxylate 95%

Supplier: AMBEED, INC
Warning

A201460-1G A201460-100MG A201460-250MG A201460-1G
103817-296EA 235 USD InStock
103817-296 76873-418 76873-420 77302-754
Methyl oxazole-2-carboxylate 95%
Methyl oxazole-2-carboxylate
Precaución: for Research Use Only


Formula: C₅H₅NO₃
Storage Temperature: Ambient
MDL Number: MFCD07699294
CAS Number: 31698-88-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty