Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl phenyl carbonate 98%

Supplier: AMBEED, INC
Warning

A189967-1G A189967-5G A189967-25G A189967-5G A189967-100G A189967-10G A189967-1G
103816-246EA 6 USD InStock
103816-246 103846-840 77084-630 77084-632 77248-542 77248-544 77302-194
Methyl phenyl carbonate 98%
Methyl phenyl carbonate
Precaución: for Research Use Only


Formula: C₈H₈O₃
Boiling Pt: 212…213 °C (760 mm Hg)
Flash Pt: 97.1 °C
Storage Temperature: Ambient
MDL Number: MFCD04039782
CAS Number: 13509-27-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty