Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(Methylsulfonyl)benzene 98%

Supplier: AMBEED, INC
Warning

A850109-5G A850109-25G A850109-500G A850109-5G A850109-100G
103841-232EA 8 USD InStock
103841-232 76845-478 77105-920 77308-452 77686-886
(Methylsulfonyl)benzene 98%
(Methylsulphonyl)benzene
Precaución: for Research Use Only


Formula: C₇H₈O₂S
Boiling Pt: 302.6±15.0 °C (760 mm Hg)
Melting Pt: 85…87 °C
Flash Pt: 171.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00014741
CAS Number: 3112-85-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty