Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl piperazine-2-carboxylate dihydrochloride 97%

Supplier: AMBEED, INC
Warning

A441150-5G A441150-1G A441150-250MG
103827-668EA 38 USD InStock
103827-668 103838-734 76842-502 77156-060
Methyl piperazine-2-carboxylate dihydrochloride 97%
Methyl piperazine-2-carboxylate dihydrochloride
Precaución: for Research Use Only


Formula: C₆H₁₄Cl₂N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD01861815
CAS Number: 122323-88-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty