Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl quinoline-6-carboxylate 97%

Supplier: AMBEED, INC
Warning

A354547-100G A354547-10G A354547-1G A354547-25G A354547-5G
103826-126EA 6 USD InStock
103826-126 103837-130 76724-506 76724-508 76840-252 76840-254 76840-256
Methyl quinoline-6-carboxylate 97%
Methyl quinoline-6-carboxylate
Precaución: for Research Use Only


Formula: C₁₁H₉NO₂
Boiling Pt: 315.1 °C (760 mm Hg)
Melting Pt: 88 °C
Flash Pt: 144.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00956766
CAS Number: 38896-30-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty