Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-2-thiophenecarboxylate 98%

Supplier: AMBEED, INC
Warning

A179114-5G A179114-25G A179114-5G A179114-100G A179114-1G
103845-736EA 13 USD InStock
103845-736 77083-184 77083-186 77111-342 77111-344
Methyl-2-thiophenecarboxylate 98%
Methyl-2-thiophenecarboxylate
Precaución: for Research Use Only


Formula: C₆H₆O₂S
MW: 142.17 g/mol
Boiling Pt: 200.2 °C (760 mm Hg)
Flash Pt: 74.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00016895
CAS Number: 5380-42-7
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty