Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 4-bromo-2-thiazole-carboxylate 95%

Supplier: AMBEED, INC
Warning

A209909-1G A209909-5G A209909-100MG A209909-250MG A209909-1G
103817-966EA 49 USD InStock
103817-966 103848-652 77012-906 77012-908 77087-016
Methyl 4-bromo-2-thiazole-carboxylate 95%
Methyl 4-bromo-2-thiazole-carboxylate
Precaución: for Research Use Only


Formula: C₅H₄BrNO₂S
Boiling Pt: 277 °C (760 mm Hg)
Flash Pt: 121.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11114545
CAS Number: 1025468-06-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty