Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methylene diacetate 95%

Supplier: AMBEED, INC
Warning

A142609-1G A142609-250MG A142609-1G A142609-5G
103803-192EA 29 USD InStock
103803-192 76719-402 77316-424 77316-426
Methylene diacetate 95%
Methylene diacetate
Precaución: for Research Use Only


Formula: C₅H₈O₄
Boiling Pt: 164.5 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD00010751
CAS Number: 628-51-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty