Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methanesulfonamide 97%

Supplier: AMBEED, INC
Warning

A124693-1000G A124693-100G A124693-25G A124693-500G A124693-5G A124693-1000G
103808-302EA 56 USD InStock
103808-302 77108-780 77108-782 77108-784 77108-786 77312-708
Methanesulfonamide 97%
Methanesulphonamide
Precaución: for Research Use Only


Formula: CH₅NO₂S
Boiling Pt: 208.2 °C (760 mm Hg)
Melting Pt: 85…89 °C
Flash Pt: 79.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00007940
CAS Number: 3144-09-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty