Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Morpholine-2-carbonitrile hydrochloride 97%

Supplier: AMBEED, INC
Danger

A823795-100MG A823795-250MG A823795-1G
103841-148EA 1407 USD InStock
103841-148 76880-400 76880-402 77308-404
Morpholine-2-carbonitrile hydrochloride 97%
Morpholine-2-carbonitrile hydrochloride
Precaución: for Research Use Only


Formula: C₅H₉ClN₂O
Storage Temperature: Ambient
MDL Number: MFCD14582597
CAS Number: 1205751-07-0
UN: 2811
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty