Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Formylmorpholine 98%

Supplier: AMBEED, INC
Warning

A263168-100G A263168-500G A263168-1000G
103809-864EA 37 USD InStock
103809-864 76722-872 76722-874 76722-876 77092-140
4-Formylmorpholine 98%
4-Formylmorpholine
Precaución: for Research Use Only


Formula: C₅H₉NO₂
Boiling Pt: 236.5 °C (760 mm Hg)
Melting Pt: 23 °C
Flash Pt: 125 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006170
CAS Number: 4394-85-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty