Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Tolylboronic acid 97%

Supplier: AMBEED, INC
Warning

A156561-5G A156561-100G A156561-10G A156561-500G A156561-5G A156561-25G
103843-102EA 5 USD InStock
103843-102 76768-372 76768-374 76768-376 76768-378 77300-196 77318-894
3-Tolylboronic acid 97%
3-Tolylboronic acid
Precaución: for Research Use Only


Formula: C₇H₉BO₂
MW: 135.96 g/mol
Boiling Pt: 290.4 °C (760 mm Hg)
Melting Pt: 165
Flash Pt: 129.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00040198
CAS Number: 17933-03-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty