Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Acetamidophthalic anhydride 97%

Supplier: AMBEED, INC
Warning

A313057-1G A313057-5G A313057-100G A313057-10G A313057-250MG A313057-1G A313057-5G
103814-714EA 8 USD InStock
103814-714 103835-904 77167-152 77167-154 77167-156 77306-076 77306-078
3-Acetamidophthalic anhydride 97%
3-Acetamidophthalic anhydride
Precaución: for Research Use Only


Formula: C₁₀H₇NO₄
MW: 205.17 g/mol
Melting Pt: 185…186 °C
Flash Pt: 249.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00453138
CAS Number: 6296-53-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty