Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-(2-Hydroxyethyl)dodecanamide 98%

Supplier: AMBEED, INC
Warning

A459808-5G A459808-100MG A459808-10G A459808-250MG A459808-1G A459808-5G A459808-100G A459808-25G
103827-904EA 21 USD InStock
103827-904 103838-980 76804-616 76804-618 76804-620 76842-854 76842-856 77880-002 77880-003
N-(2-Hydroxyethyl)dodecanamide 98%
N-(2-Hydroxyethyl)dodecanamide
Precaución: for Research Use Only


Formula: C₁₄H₂₉NO₂
Boiling Pt: 410.7 °C (760 mm Hg)
Melting Pt: 88…89 °C
Storage Temperature: Ambient
MDL Number: MFCD00020561
CAS Number: 142-78-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty