Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(2-Hydroxyethyl)piperazine 97%

Supplier: AMBEED, INC
Warning

A101110-1000G A101110-100G A101110-25G A101110-500G
103807-780EA 40 USD InStock
103807-780 77025-164 77025-166 77025-168
1-(2-Hydroxyethyl)piperazine 97%
1-(2-Hydroxyethyl)piperazine
Precaución: for Research Use Only


Formula: C₆H₁₄N₂O
MW: 130.19 g/mol
Boiling Pt: 246
Melting Pt: -38
Flash Pt: 135 °C
Storage Temperature: Ambient
MDL Number: MFCD00005970
CAS Number: 103-76-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty