Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl)acetamide 98%

Supplier: AMBEED, INC
Warning

A189941-1G A189941-5G A189941-100MG A189941-250MG A189941-1G A189941-5G
103816-238EA 51 USD InStock
103816-238 103846-834 76974-816 76974-818 77084-612 77084-614
N-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl)acetamide 98%
N-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl)acetamide
Precaución: for Research Use Only


Formula: C₁₃H₁₉BN₂O₃
Boiling Pt: 451.8±30.0 °C (760 mm Hg)
Flash Pt: 227 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11878181
CAS Number: 1220220-21-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty