Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-[4-Cyano-3-(trifluoromethyl)phenyl]-2-methyloxirane-2-carboxamide 98%

Supplier: AMBEED, INC
Warning

A173960-5G A173960-100G A173960-25G A173960-5G
103845-192EA 7 USD InStock
103845-192 76872-518 77082-478 77082-480
N-[4-Cyano-3-(trifluoromethyl)phenyl]-2-methyloxirane-2-carboxamide 98%
N-[4-Cyano-3-(trifluoromethyl)phenyl]-2-methyloxirane-2-carboxamide
Precaución: for Research Use Only


Formula: C₁₂H₉F₃N₂O₂
MW: 270.21 g/mol
Boiling Pt: 436.4 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD08460201
CAS Number: 90357-51-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty