Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-Methyl-p-toluenesulfonamide 97%

Supplier: AMBEED, INC
Warning

A162891-1000G A162891-100G A162891-10G A162891-500G A162891-25G A162891-1000G
103808-918EA 49 USD InStock
103808-918 76720-202 76720-204 76720-206 77300-616 77319-996
N-Methyl-p-toluenesulfonamide 97%
N-Methyl-p-toluenesulfonamide
Precaución: for Research Use Only


Formula: C₈H₁₁NO₂S
Boiling Pt: 296.5 °C (760 mm Hg)
Melting Pt: 76…79 °C
Flash Pt: 133.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00008285
CAS Number: 640-61-9
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty