Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N,N-Dimethyl-1-(morpholin-2-yl)methanamine 97%

Supplier: AMBEED, INC
Warning

A252954-1G A252954-5G A252954-10G A252954-250MG A252954-1G A252954-5G A252954-100MG
103812-066EA 112 USD InStock
103812-066 103851-914 77000-724 77000-726 77091-338 77304-594 77881-682
N,N-Dimethyl-1-(morpholin-2-yl)methanamine 97%
N,N-Dimethyl-1-(morpholin-2-yl)methanamine
Precaución: for Research Use Only


Formula: C₇H₁₆N₂O
MW: 144.2147 g/mol
Boiling Pt: 199 °C (760 mm Hg)
Flash Pt: 74 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03990577
CAS Number: 122894-56-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty