Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N,N-Dimethyl-2-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazol-1-yl)ethanamine 95%

Supplier: AMBEED, INC
Warning

A227581-1G A227581-5G A227581-100MG A227581-250MG A227581-1G A227581-10G A227581-25G
103819-326EA 80 USD InStock
103819-326 103850-232 76975-992 76975-994 77089-004 77881-580 77881-581
N,N-Dimethyl-2-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazol-1-yl)ethanamine 95%
N,N-Dimethyl-2-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazol-1-yl)ethanamine
Precaución: for Research Use Only


Formula: C₁₃H₂₄BN₃O₂
Boiling Pt: 365.765 °C (760 mm Hg)
Flash Pt: 175.009 °C
Storage Temperature: Refrigerator
MDL Number: MFCD16660235
CAS Number: 877149-80-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty