Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-[2-(tert-Butyldimethylsilyloxy)ethyl]methylamine 97%

Supplier: AMBEED, INC
Warning

A166317-1G A166317-5G A166317-10G A166317-1G A166317-5G
103806-050EA 27 USD InStock
103806-050 103844-312 77096-332 77300-826 77300-828
N-[2-(tert-Butyldimethylsilyloxy)ethyl]methylamine 97%
N-[2-(tert-Butyldimethylsilyloxy)ethyl]methylamine
Precaución: for Research Use Only


Formula: C₉H₂₃NOSi
Storage Temperature: Refrigerator
MDL Number: MFCD20662141
CAS Number: 204580-28-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty