Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,1,4,7,10,10-Hexamethyltriethylenetetramine 97%

Supplier: AMBEED, INC
Danger

A407765-1G A407765-5G A407765-1G A407765-25G A407765-5G
103827-160EA 17 USD InStock
103827-160 103838-202 77307-112 77307-114 77307-116
1,1,4,7,10,10-Hexamethyltriethylenetetramine 97%
1,1,4,7,10,10-Hexamethyltriethylenetetramine
Precaución: for Research Use Only


Formula: C₁₂H₃₀N₄
Boiling Pt: 130 °C (11 mm Hg)
Flash Pt: 102 °C
Storage Temperature: Ambient
MDL Number: MFCD00025684
CAS Number: 3083-10-1
UN: 2735
ADR: 8,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty