Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,1'-Azobis(N,N-dimethylformamide) 97%

Supplier: AMBEED, INC
Danger

A181208-1G A181208-5G A181208-1G A181208-25G A181208-10G A181208-250MG A181208-5G
103807-588EA 9 USD InStock
103807-588 103845-954 77083-476 77083-478 77151-614 77151-616 77301-708
1,1'-Azobis(N,N-dimethylformamide) 97%
1,1'-Azobis(N,N-dimethylformamide)
Precaución: for Research Use Only


Formula: C₆H₁₂N₄O₂
Boiling Pt: 220.4 °C (760 mm Hg)
Melting Pt: 112 °C
Flash Pt: 87.1 °C
Storage Temperature: Freezer
MDL Number: MFCD00008318
CAS Number: 10465-78-8
UN: 1325
ADR: 4.1,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty