Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N6-Methyladenosine 97%

Supplier: AMBEED, INC
Warning

A170736-1G A170736-5G A170736-1G A170736-5G A170736-100MG A170736-1MG
103799-418EA 13 USD InStock
103799-418 103806-534 103844-808 77081-922 77081-924 77163-562 77163-564
N6-Methyladenosine 97%
N6-Methyladenosine
Precaución: for Research Use Only


Formula: C₁₁H₁₅N₅O₄
MW: 281.26 g/mol
Boiling Pt: 649.1 °C (760 mm Hg)
Flash Pt: 346.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005739
CAS Number: 1867-73-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty