Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Naftifine hydrochloride 98%

Supplier: AMBEED, INC
Warning

A287696-1G A287696-5G A287696-1MG A287696-25G
103799-568EA 38 USD InStock
103799-568 103813-770 103834-912 76723-418 77879-560
Naftifine hydrochloride 98%
Naftifine hydrochloride
Precaución: for Research Use Only


Formula: C₂₁H₂₁N
Melting Pt: 172…175 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00059047
CAS Number: 65473-14-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty