Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Boc-2-piperidone 98%

Supplier: AMBEED, INC
Warning

A115430-1G A115430-5G A115430-100G A115430-10G A115430-25G A115430-500G A115430-1G A115430-5G
103798-560EA 5 USD InStock
103798-560 103824-416 77108-240 77108-242 77108-244 77108-246 77311-234 77311-236
1-Boc-2-piperidone 98%
1-Boc-2-piperidone
Precaución: for Research Use Only


Formula: C₁₀H₁₇NO₃
MW: 199.25 g/mol
Melting Pt: 29…36 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD02179046
CAS Number: 85908-96-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty