Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl-3-oxoazepane-1-carboxylate 95%

Supplier: AMBEED, INC
Warning

A575845-1G A575845-250MG A575845-5G
103828-838EA 35 USD InStock
103828-838 103839-988 76819-216 76844-126
tert-Butyl-3-oxoazepane-1-carboxylate 95%
tert-Butyl-3-oxoazepane-1-carboxylate
Precaución: for Research Use Only


Formula: C₁₁H₁₉NO₃
Boiling Pt: 308.2 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD08443803
CAS Number: 870842-23-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty