Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-Boc-4-piperidineethanol 97%

Supplier: AMBEED, INC
Warning

A150552-1G A150552-5G A150552-100G A150552-10G A150552-500G A150552-1G A150552-25G A150552-5G
103804-182EA 6 USD InStock
103804-182 103842-318 76871-610 76871-612 76871-614 77317-840 77317-842 77317-844
N-Boc-4-piperidineethanol 97%
N-Boc-4-piperidineethanol
Precaución: for Research Use Only


Formula: C₁₂H₂₃NO₃
MW: 229.3159 g/mol
Boiling Pt: 324 °C (760 mm Hg)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD03427086
CAS Number: 89151-44-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty