Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Nɑ-Carbobenzoxy-D-asparagine 97%

Supplier: AMBEED, INC
Warning

A200827-1G A200827-5G A200827-500G A200827-10G
103817-242EA 5 USD InStock
103817-242 103847-876 76975-256 77881-472
Nɑ-Carbobenzoxy-D-asparagine 97%
Nalpha-Carbobenzoxy-D-asparagine
Precaución: for Research Use Only


Formula: C₁₂H₁₄N₂O₅
MW: 266.25 g/mol
Boiling Pt: 580.6±50.0 °C (760 mm Hg)
Melting Pt: 162…164 °C
Flash Pt: 304.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00065696
CAS Number: 4474-86-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty