Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N'-Hydroxyacetimidamide 98%

Supplier: AMBEED, INC
Danger

A371718-1G A371718-100G A371718-10G A371718-25G A371718-5G A371718-1G
103826-490EA 8 USD InStock
103826-490 103837-508 76694-338 76694-340 76840-758 76840-760 77306-840
N'-Hydroxyacetimidamide 98%
N'-Hydroxyacetimidamide
Precaución: for Research Use Only


Formula: C₂H₆N₂O
Boiling Pt: 197.9 °C (760 mm Hg)
Melting Pt: 134 °C
Storage Temperature: Ambient
MDL Number: MFCD09859749
CAS Number: 22059-22-9
UN: 3077
ADR: 9,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty