Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-Methoxy-N-methylacetamide 98%

Supplier: AMBEED, INC
Danger

A310673-5G A310673-10G A310673-25G A310673-5G
103835-814EA 11 USD InStock
103835-814 76723-826 76838-586 77306-034
N-Methoxy-N-methylacetamide 98%
N-Methoxy-N-methylacetamide
Precaución: for Research Use Only


Formula: C₄H₉NO₂
Boiling Pt: 152 °C (760 mm Hg)
Flash Pt: 49 °C
Storage Temperature: Ambient
MDL Number: MFCD00060098
CAS Number: 78191-00-1
UN: 1993
ADR: 3,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty