Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-Methoxy-N-methylbenzamide 98%

Supplier: AMBEED, INC
Warning

A170473-1G A170473-5G A170473-100G A170473-10G A170473-1G A170473-25G A170473-5G
103806-504EA 5 USD InStock
103806-504 103844-778 76720-506 76720-508 77301-058 77301-060 77301-062
N-Methoxy-N-methylbenzamide 98%
N-Methoxy-N-methylbenzamide
Precaución: for Research Use Only


Formula: C₉H₁₁NO₂
MW: 165.19 g/mol
Boiling Pt: 288.2 °C (760 mm Hg)
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00075320
CAS Number: 6919-61-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty