Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-Methoxy-N-methylcyclopropanecarboxamide 97%

Supplier: AMBEED, INC
Warning

A130474-1G A130474-100G A130474-1G A130474-25G A130474-5G
103801-430EA 9 USD InStock
103801-430 76870-696 77313-864 77313-866 77313-868
N-Methoxy-N-methylcyclopropanecarboxamide 97%
N-Methoxy-N-methylcyclopropanecarboxamide
Precaución: For research use only.


Formula: C₆H₁₁NO₂
Boiling Pt: 151.5±23.0 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD11845572
CAS Number: 147356-78-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty