Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-Methyl-4-bromobenzylamine Hydrochloride 97%

Supplier: AMBEED, INC
Warning

A100238-1G A100238-5G A100238-1G A100238-5G
103795-314EA 14 USD InStock
103795-314 103821-024 77296-604 77296-608
N-Methyl-4-bromobenzylamine Hydrochloride 97%
N-Methyl-4-bromobenzylamine Hydrochloride
Precaución: for Research Use Only


Formula: C₈H₁₁BrClN
MW: 236.54 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07106760
CAS Number: 874-73-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty