Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Non-8-yn-1-ol 97%

Supplier: AMBEED, INC
Warning

A162678-1G A162678-5G A162678-100G A162678-250MG A162678-1G A162678-25G A162678-5G
103805-648EA 20 USD InStock
103805-648 103843-898 76689-876 76689-878 77319-966 77319-968 77319-970
Non-8-yn-1-ol 97%
Non-8-yn-1-ol
Precaución: for Research Use Only


Formula: C₉H₁₆O
Storage Temperature: Refrigerator
MDL Number: MFCD00967780
CAS Number: 10160-28-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty