Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

n-Pentylboronic acid 98%

Supplier: AMBEED, INC
Warning

A143829-1G A143829-10G A143829-1G A143829-25G A143829-5G
103803-322EA 9 USD InStock
103803-322 77272-594 77316-622 77316-624 77316-626
n-Pentylboronic acid 98%
n-Pentylboronic acid
Precaución: for Research Use Only


Formula: C₅H₁₃BO₂
Boiling Pt: 207.9 °C (760 mm Hg)
Melting Pt: 90…94 °C
Storage Temperature: Freezer
MDL Number: MFCD01074651
CAS Number: 4737-50-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty