Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Amino-N-tert-butylbenzenesulfonamide 98%

Supplier: AMBEED, INC
Warning

A107312-1G A107312-5G A107312-250MG A107312-1G A107312-5G A107312-25G
103797-168EA 18 USD InStock
103797-168 103822-936 77107-746 77283-574 77283-576 77684-969
3-Amino-N-tert-butylbenzenesulfonamide 98%
3-Amino-N-tert-butylbenzenesulphonamide
Precaución: for Research Use Only


Formula: C₁₀H₁₆N₂O₂S
Boiling Pt: 387.3 °C (760 mm Hg)
Flash Pt: 188 °C
Storage Temperature: Ambient
MDL Number: MFCD06660528
CAS Number: 608523-94-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty