Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Cyclopropyl methoxylamine hydrochloride 97%

Supplier: AMBEED, INC
Warning

A256003-1G A256003-5G A256003-250MG A256003-1G
103812-228EA 54 USD InStock
103812-228 103852-088 76692-626 77304-662
Cyclopropyl methoxylamine hydrochloride 97%
Cyclopropyl methoxylamine hydrochloride
Precaución: for Research Use Only


Formula: C₄H₉NO·ClH
MW: 123.581 g/mol
Melting Pt: 170…172 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08689925
CAS Number: 74124-04-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty