Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Octen-3-one 95%

Supplier: AMBEED, INC
Danger

A193991-1G A193991-5G A193991-1G A193991-5G A193991-10G A193991-25G
103816-610EA 10 USD InStock
103816-610 103847-218 77085-126 77085-128 77236-990 77302-378
1-Octen-3-one 95%
1-Octen-3-one
Precaución: for Research Use Only


Formula: C₈H₁₄O
MW: 126.20 g/mol
Boiling Pt: 166…167 °C (760 mm Hg)
Flash Pt: 73.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00036558
CAS Number: 4312-99-6
UN: 1224
ADR: 3 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty