Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,18-Octadecanedioic acid 98%

Supplier: AMBEED, INC
Warning

A182546-1G A182546-5G A182546-10G A182546-1G A182546-25G A182546-5G
103807-724EA 7 USD InStock
103807-724 103846-098 76720-938 77083-644 77083-646 77083-648
1,18-Octadecanedioic acid 98%
1,18-Octadecanedioic acid
Precaución: for Research Use Only


Formula: C₁₈H₃₄O₄
Boiling Pt: 480.9 °C (760 mm Hg)
Melting Pt: 125 °C
Flash Pt: 258.8 °C
Storage Temperature: Ambient
MDL Number: MFCD00142369
CAS Number: 871-70-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty