Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

o-Tolylacetic acid 98%

Supplier: AMBEED, INC
Warning

A171202-1000G A171202-25G A171202-100G A171202-500G A171202-5G
103809-036EA 92 USD InStock
103809-036 77082-010 77082-012 77248-010 77248-012 77248-014
o-Tolylacetic acid 98%
o-Tolylacetic acid
Precaución: for Research Use Only


Formula: (C₆H₄CH₃)CH₂COOH
Boiling Pt: 279.3 °C (760 mm Hg)
Melting Pt: 88…90 °C
Flash Pt: 176.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00004328
CAS Number: 644-36-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty