Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Paradol 98%

Supplier: AMBEED, INC
Warning

A357502-5MG A357502-10MG A357502-50MG A357502-1MG A357502-100MG A357502-1G
103791-654EA 31 USD InStock
103791-654 103792-662 103795-144 103799-620 77240-092 77616-570 77746-706
Paradol 98%
Paradol
Precaución: for Research Use Only


Formula: C₁₇H₂₆O₃
Boiling Pt: 406.5±30.0 °C (760 mm Hg)
Flash Pt: 140 °C
Storage Temperature: Ambient
MDL Number: MFCD01736103
CAS Number: 27113-22-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty