Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Pentaerythritol tris(3-(1-aziridinyl)propionate) 99% Solid content 99%

Supplier: AMBEED, INC
Warning

A352994-5G A352994-100G A352994-10G A352994-1000G
103810-206EA 206 USD InStock
103810-206 103837-082 76828-614 76828-616 77306-638
Pentaerythritol tris(3-(1-aziridinyl)propionate) 99% Solid content 99%
Pentaerythritol tris(3-(1-aziridinyl)propionate)
Precaución: for Research Use Only


Formula: C₂₀H₃₃N₃O₇
Boiling Pt: 542.3 °C (760 mm Hg)
Flash Pt: 281.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01757346
CAS Number: 57116-45-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty