Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Pentafluorophenyl trifluoroacetate 98%

Supplier: AMBEED, INC
Danger

A314355-5G A314355-10G A314355-1G A314355-25G A314355-5G
103825-032EA 14 USD InStock
103825-032 103835-952 76723-854 76838-762 76838-764 77306-112
Pentafluorophenyl trifluoroacetate 98%
Pentafluorophenyl trifluoroacetate
Precaución: for Research Use Only


Formula: C₈F₈O₂
MW: 280.07 g/mol
Boiling Pt: 122.5 °C (760 mm Hg)
Flash Pt: 52 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00134438
CAS Number: 14533-84-7
UN: 3272
ADR: 3 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty